Learn More
4-Cyano-L-phenylalanine, 95%
CAS: 167479-78-9 | C10H10N2O2 | 190.202 g/mol
$99.74 - $939.14
Chemical Identifiers
| CAS | 167479-78-9 |
|---|---|
| Molecular Formula | C10H10N2O2 |
| Molecular Weight (g/mol) | 190.202 |
| MDL Number | MFCD00270363 |
| InChI Key | KWIPUXXIFQQMKN-VIFPVBQESA-N |
| Synonym | l-4-cyanophenylalanine, 4-cyano-l-phenylalanine, s-2-amino-3-4-cyanophenyl propanoic acid, h-phe 4-cn-oh, 2s-2-amino-3-4-cyanophenyl propanoic acid, 4-cyanophenylalanine, s-2-amino-3-4-cyanophenyl propionic acid, d-4-cn-phe-oh |
| PubChem CID | 1501865 |
| IUPAC Name | (2S)-2-amino-3-(4-cyanophenyl)propanoic acid |
| SMILES | C1=CC(=CC=C1CC(C(=O)O)N)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH63572MD
|
Thermo Scientific Chemicals
H63572MD |
250 mg |
Each for $99.74
|
|
|||||
|
AAH6357203
|
Thermo Scientific Chemicals
H6357203 |
1 g |
Each for $252.44
|
|
|||||
|
AAH6357206
|
Thermo Scientific Chemicals
H6357206 |
5 g |
Each for $939.14
|
|
|||||
Description
4-Cyano-L-phenylalanine used as a intermediate in organic synthesis, pharmaceutical, chemical research and biochemical application.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Cyano-L-phenylalanine used as a intermediate in organic synthesis, pharmaceutical, chemical research and biochemical application.
Solubility
Slightly soluble in water.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
| 167479-78-9 | |
| 190.202 | |
| KWIPUXXIFQQMKN-VIFPVBQESA-N | |
| 1501865 | |
| C1=CC(=CC=C1CC(C(=O)O)N)C#N |
| C10H10N2O2 | |
| MFCD00270363 | |
| l-4-cyanophenylalanine, 4-cyano-l-phenylalanine, s-2-amino-3-4-cyanophenyl propanoic acid, h-phe 4-cn-oh, 2s-2-amino-3-4-cyanophenyl propanoic acid, 4-cyanophenylalanine, s-2-amino-3-4-cyanophenyl propionic acid, d-4-cn-phe-oh | |
| (2S)-2-amino-3-(4-cyanophenyl)propanoic acid |
Specifications
| 167479-78-9 | |
| MFCD00270363 | |
| l-4-cyanophenylalanine, 4-cyano-l-phenylalanine, s-2-amino-3-4-cyanophenyl propanoic acid, h-phe 4-cn-oh, 2s-2-amino-3-4-cyanophenyl propanoic acid, 4-cyanophenylalanine, s-2-amino-3-4-cyanophenyl propionic acid, d-4-cn-phe-oh | |
| KWIPUXXIFQQMKN-VIFPVBQESA-N | |
| (2S)-2-amino-3-(4-cyanophenyl)propanoic acid | |
| 190.202 | |
| 190.2 | |
| Solid |
| C10H10N2O2 | |
| UN3439 | |
| Slightly soluble in water. | |
| C1=CC(=CC=C1CC(C(=O)O)N)C#N | |
| 250 mg | |
| 1501865 | |
| 98% | |
| 4-Cyano-L-phenylalanine |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only