Learn More
4-Cyano-L-phenylalanine, 95%
CAS: 167479-78-9 | C10H10N2O2 | 190.202 g/mol
Supplier: Thermo Scientific Chemicals H6357203
Description
4-Cyano-L-phenylalanine used as a intermediate in organic synthesis, pharmaceutical, chemical research and biochemical application.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Cyano-L-phenylalanine used as a intermediate in organic synthesis, pharmaceutical, chemical research and biochemical application.
Solubility
Slightly soluble in water.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications
| 4-Cyano-L-phenylalanine | |
| C10H10N2O2 | |
| UN3439 | |
| Slightly soluble in water. | |
| C1=CC(=CC=C1CC(C(=O)O)N)C#N | |
| 1 g | |
| 1501865 | |
| 98% |
| 167479-78-9 | |
| MFCD00270363 | |
| l-4-cyanophenylalanine, 4-cyano-l-phenylalanine, s-2-amino-3-4-cyanophenyl propanoic acid, h-phe 4-cn-oh, 2s-2-amino-3-4-cyanophenyl propanoic acid, 4-cyanophenylalanine, s-2-amino-3-4-cyanophenyl propionic acid, d-4-cn-phe-oh | |
| KWIPUXXIFQQMKN-VIFPVBQESA-N | |
| (2S)-2-amino-3-(4-cyanophenyl)propanoic acid | |
| 190.202 | |
| 190.2 | |
| Solid |
Chemical Identifiers
| 167479-78-9 | |
| 190.202 | |
| KWIPUXXIFQQMKN-VIFPVBQESA-N | |
| 1501865 | |
| C1=CC(=CC=C1CC(C(=O)O)N)C#N |
| C10H10N2O2 | |
| MFCD00270363 | |
| l-4-cyanophenylalanine, 4-cyano-l-phenylalanine, s-2-amino-3-4-cyanophenyl propanoic acid, h-phe 4-cn-oh, 2s-2-amino-3-4-cyanophenyl propanoic acid, 4-cyanophenylalanine, s-2-amino-3-4-cyanophenyl propionic acid, d-4-cn-phe-oh | |
| (2S)-2-amino-3-(4-cyanophenyl)propanoic acid |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only