missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyano-4'-propylbiphenyl 99.0+%, TCI America™
Supplier: TCI America C27645G
Specifications
| 4-Cyano-4′-propylbiphenyl | |
| 67°C | |
| C16H15N | |
| 5 g | |
| XFMPTZWVMVMELB-UHFFFAOYSA-N | |
| 4'-propyl-[1,1'-biphenyl]-4-carbonitrile | |
| 93891 | |
| ≥99.0% (GC) |
| 58743-76-3 | |
| White | |
| MFCD00505987 | |
| 4'-propyl-1,1'-biphenyl-4-carbonitrile, 4-propyl-4'-cyanobiphenyl, 4-cyano-4'-propylbiphenyl, 1,1'-biphenyl-4-carbonitrile, 4'-propyl, 4-cyano-4'-n-propylbiphenyl, 4-4-propylphenyl benzonitrile, 4-cyano-4'-n-propyl-biphenyl, 4'-propyl-4-cyanobiphenyl, 4'-propylbiphenyl-4-carbonitrile, 4-trans-4-propylphenyl benzonitrile | |
| CCCC1=CC=C(C=C1)C1=CC=C(C=C1)C#N | |
| 221.30 | |
| 221.30 | |
| Crystalline Powder |
Chemical Identifiers
| 58743-76-3 | |
| 221.30 | |
| XFMPTZWVMVMELB-UHFFFAOYSA-N | |
| 93891 | |
| CCCC1=CC=C(C=C1)C1=CC=C(C=C1)C#N |
| C16H15N | |
| MFCD00505987 | |
| 4'-propyl-1,1'-biphenyl-4-carbonitrile, 4-propyl-4'-cyanobiphenyl, 4-cyano-4'-propylbiphenyl, 1,1'-biphenyl-4-carbonitrile, 4'-propyl, 4-cyano-4'-n-propylbiphenyl, 4-4-propylphenyl benzonitrile, 4-cyano-4'-n-propyl-biphenyl, 4'-propyl-4-cyanobiphenyl, 4'-propylbiphenyl-4-carbonitrile, 4-trans-4-propylphenyl benzonitrile | |
| 4'-propyl-[1,1'-biphenyl]-4-carbonitrile |
Safety and Handling
TSCA : Yes