missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4'-propylbiphenyl 99.0+%, TCI America™

Catalog No. C27641G
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
C27641G 1 g
C27645G 5 g
2 options

Catalog No. C27641G

Supplier: TCI America C27641G

Only null left

Chemical Identifiers

CAS 58743-76-3
Molecular Formula C16H15N
Molecular Weight (g/mol) 221.30
MDL Number MFCD00505987
InChI Key XFMPTZWVMVMELB-UHFFFAOYSA-N
Synonym 4'-propyl-1,1'-biphenyl-4-carbonitrile, 4-propyl-4'-cyanobiphenyl, 4-cyano-4'-propylbiphenyl, 1,1'-biphenyl-4-carbonitrile, 4'-propyl, 4-cyano-4'-n-propylbiphenyl, 4-4-propylphenyl benzonitrile, 4-cyano-4'-n-propyl-biphenyl, 4'-propyl-4-cyanobiphenyl, 4'-propylbiphenyl-4-carbonitrile, 4-trans-4-propylphenyl benzonitrile
PubChem CID 93891
IUPAC Name 4'-propyl-[1,1'-biphenyl]-4-carbonitrile
SMILES CCCC1=CC=C(C=C1)C1=CC=C(C=C1)C#N

Specifications

Melting Point 67°C
Color White
Quantity 1 g
Formula Weight 221.30
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-4′-propylbiphenyl