missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4″-propyl-p-terphenyl 98.0+%, TCI America™

Catalog No. C32711G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C32711G Supplier TCI America Supplier No. C32711G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 54296-25-2
Molecular Formula C22H19N
Molecular Weight (g/mol) 297.40
MDL Number MFCD00799422
InChI Key PWZKBBQJWUEPOK-UHFFFAOYSA-N
Synonym 4′C′C-Propyl-[1,1′C:4′C,1′C′C-terphenyl]-4-carbonitrile
PubChem CID 14439274
IUPAC Name 4'-(4-propylphenyl)-[1,1'-biphenyl]-4-carbonitrile
SMILES CCCC1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C#N

Specifications

Color White
Quantity 1 g
Formula Weight 297.40
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-4′′-propyl-p-terphenyl