missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4'-propoxy-1,1'-biphenyl 98.0+%, TCI America™

Catalog No. C29111G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29111G 1 g
C29115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29111G Supplier TCI America Supplier No. C29111G
Only null left

Chemical Identifiers

CAS 52709-86-1
Molecular Formula C16H15NO
Molecular Weight (g/mol) 237.302
MDL Number MFCD00183206
InChI Key RZCQJZFWJZHMPK-UHFFFAOYSA-N
Synonym 4'-propoxy-1,1'-biphenyl-4-carbonitrile, 3ocb, 1,1'-biphenyl-4-carbonitrile, 4'-propoxy, 4-propoxy-4'-cyanobiphenyl, 4-cyano-4'-propoxybiphenyl, 4-propoxycyanodiphenyl, 4-propoxy-1,1'-biphenyl-4'-carbonitrile, 4-4-propoxyphenyl benzonitrile, 4'-propoxybiphenyl-4-carbonitrile, 4'-propoxy-4-cyanobiphenyl
PubChem CID 104291
IUPAC Name 4-(4-propoxyphenyl)benzonitrile
SMILES CCCOC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 73°C
Color White
Quantity 1 g
Formula Weight 237.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-4′-propoxy-1,1′-biphenyl
RTECSNumber DV1764600
TSCA Yes