missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4'-pentyloxybiphenyl 98.0+%, TCI America™

Catalog No. p-7125055
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15511G 1 g
C15515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15511G Supplier TCI America Supplier No. C15511G

Chemical Identifiers

CAS 52364-71-3
Molecular Formula C18H19NO
Molecular Weight (g/mol) 265.356
MDL Number MFCD00074879
InChI Key RDISTOCQRJJICR-UHFFFAOYSA-N
Synonym 4'-pentyloxy-1,1'-biphenyl-4-carbonitrile, 4-cyano-4'-pentyloxybiphenyl, 4-pentyloxy-4'-cyanobiphenyl, 4-amyloxycyanodiphenyl, 5ocb, 1,1'-biphenyl-4-carbonitrile, 4'-pentyloxy, 4'-pentyloxy biphenyl-4-carbonitrile, 5cob, m 15 liquid crystal, nzh k 1
PubChem CID 104171
IUPAC Name 4-(4-pentoxyphenyl)benzonitrile
SMILES CCCCCOC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 52°C
Color White
Quantity 1 g
Formula Weight 265.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-4′-pentyloxybiphenyl
RTECSNumber DV1764550
TSCA Yes