missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4'-pentylbiphenyl 98.0+%, TCI America™

Catalog No. p-7125054
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15501G 1 g
C15505G 5 g
C155025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C15501G Supplier TCI America Supplier No. C15501G

Chemical Identifiers

CAS 40817-08-1
Molecular Formula C18H19N
Molecular Weight (g/mol) 249.357
MDL Number MFCD00036350
InChI Key HHPCNRKYVYWYAU-UHFFFAOYSA-N
Synonym 4-cyano-4'-pentylbiphenyl, 4'-pentyl-1,1'-biphenyl-4-carbonitrile, 4-cyano-4'-n-pentylbiphenyl, 1,1'-biphenyl-4-carbonitrile, 4'-pentyl, 4'-pentyl-4-biphenylcarbonitrile, 4-pentyl-4'-cyanobiphenyl, 4'-amyl-4-biphenylcarbonitrile, 4'-pentylbiphenyl-4-carbonitrile, 4-amyl-4'-cyanobiphenyl, 4-n-pentyl-4'-cyanobiphenyl
PubChem CID 92319
IUPAC Name 4-(4-pentylphenyl)benzonitrile
SMILES CCCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Color White
Boiling Point 215°C
Quantity 1 g
Formula Weight 249.36
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Cyano-4′-pentylbiphenyl