missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4″-pentyl-p-terphenyl 98.0+%, TCI America™

Catalog No. p-7125717
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29101G 1 g
C29105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29101G Supplier TCI America Supplier No. C29101G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 54211-46-0
Molecular Formula C24H23N
Molecular Weight (g/mol) 325.455
MDL Number MFCD00799423
InChI Key AITQOXOBSMXBRV-UHFFFAOYSA-N
Synonym 4-pentyl-1,1':4',1-terphenyl-4-carbonitrile, 4-cyano-4-n-pentyl-p-terphenyl, 4-cyano-4-pentyl-p-terphenyl, 1,1':4',1-terphenyl-4-carbonitrile, 4-pentyl, 4-4-4-pentylphenyl phenyl benzonitrile, pubchem9116, 4-pentyl-p-terphenyl-4-carbonitrile, 4-cyano-4'-pentyl-p-terphenyl
PubChem CID 104707
IUPAC Name 4-[4-(4-pentylphenyl)phenyl]benzonitrile
SMILES CCCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)C#N

Specifications

Melting Point 130°C
Color White
Quantity 1 g
Formula Weight 325.46
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-4′′-pentyl-p-terphenyl