missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4'-nonylbiphenyl 98.0+%, TCI America™

Catalog No. C32681G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C32681G Supplier TCI America Supplier No. C32681G

Chemical Identifiers

CAS 52709-85-0
Molecular Formula C22H27N
Molecular Weight (g/mol) 305.47
MDL Number MFCD00482592
InChI Key REDSMJNHKIHBTE-UHFFFAOYSA-N
Synonym 4′C-Nonyl-4-biphenylcarbonitrile, 4′C-Nonyl-[1,1′C-biphenyl]-4-carbonitrile, 9CB
PubChem CID 104290
IUPAC Name 4'-nonyl-[1,1'-biphenyl]-4-carbonitrile
SMILES CCCCCCCCCC1=CC=C(C=C1)C1=CC=C(C=C1)C#N

Specifications

Melting Point 42°C
Color White
Boiling Point 220°C
Quantity 1 g
Formula Weight 305.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-4′-nonylbiphenyl
TSCA Yes
Recommended Storage Refrigerator