missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4'-n-pentyloxybiphenyl, 99%

Catalog No. p-7026471
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2247703 1 g
AAB2247706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2247703 Supplier Thermo Scientific Chemicals Supplier No. B2247703
Only null left

CAS: 52364-71-3 | C18H19NO | 265.356 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52364-71-3
Molecular Formula C18H19NO
Molecular Weight (g/mol) 265.356
MDL Number MFCD00074879
InChI Key RDISTOCQRJJICR-UHFFFAOYSA-N
Synonym 4'-pentyloxy-1,1'-biphenyl-4-carbonitrile, 4-cyano-4'-pentyloxybiphenyl, 4-pentyloxy-4'-cyanobiphenyl, 4-amyloxycyanodiphenyl, 5ocb, 1,1'-biphenyl-4-carbonitrile, 4'-pentyloxy, 4'-pentyloxy biphenyl-4-carbonitrile, 5cob, m 15 liquid crystal, nzh k 1
PubChem CID 104171
IUPAC Name 4-(4-pentoxyphenyl)benzonitrile
SMILES CCCCCOC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 48° cloudy
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN3439
Formula Weight 281.42
Percent Purity 99%
Chemical Name or Material 4-Cyano-4'-n-pentyloxybiphenyl
Hazard Category H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 257-875-9
RTECSNumber DV1764550
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only