missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4'-n-pentylbiphenyl, 99%

Catalog No. AAB2185603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2185603 1 g
AAB2185606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2185603 Supplier Thermo Scientific Chemicals Supplier No. B2185603

CAS: 40817-08-1 | C18H19N | 249.357 g/mol

4-Cyano-4'-n-pentylbiphenyl is used in the preparation of 4'-pentylbiphenyl-4-carboxylic acid by acidic hydrolysis using sulfuric acid as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyano-4′-n-pentylbiphenyl is used in the preparation of 4′-pentylbiphenyl-4-carboxylic acid by acidic hydrolysis using sulfuric acid as a reagent.

Solubility
Miscible with Acetone, chloroform, toluene, acetonitrile, methylene chloride. Immiscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 40817-08-1
Molecular Formula C18H19N
Molecular Weight (g/mol) 249.357
MDL Number MFCD00036350
InChI Key HHPCNRKYVYWYAU-UHFFFAOYSA-N
Synonym 4-cyano-4'-pentylbiphenyl, 4'-pentyl-1,1'-biphenyl-4-carbonitrile, 4-cyano-4'-n-pentylbiphenyl, 1,1'-biphenyl-4-carbonitrile, 4'-pentyl, 4'-pentyl-4-biphenylcarbonitrile, 4-pentyl-4'-cyanobiphenyl, 4'-amyl-4-biphenylcarbonitrile, 4'-pentylbiphenyl-4-carbonitrile, 4-amyl-4'-cyanobiphenyl, 4-n-pentyl-4'-cyanobiphenyl
PubChem CID 92319
IUPAC Name 4-(4-pentylphenyl)benzonitrile
SMILES CCCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 22°C to 23°C
Density 1.008
Boiling Point 212°C to 215°C (3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.533
Quantity 1 g
UN Number UN3276
Solubility Information Miscible with Acetone,chloroform,toluene,acetonitrile,methylene chloride. Immiscible with water.
Formula Weight 249.36
Percent Purity 99%
Chemical Name or Material 4-Cyano-4'-n-pentylbiphenyl
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 255-093-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only