missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4″-n-pentyl-p-terphenyl, 99%

Catalog No. AAB2260702
Change view
Click to view available options
Quantity:
0.5 g
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2260702 0.5 g
AAB2260788 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2260702 Supplier Thermo Scientific Chemicals Supplier No. B2260702
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 54211-46-0 | C24H23N | 325.455 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54211-46-0
Molecular Formula C24H23N
Molecular Weight (g/mol) 325.455
MDL Number MFCD00799423
InChI Key AITQOXOBSMXBRV-UHFFFAOYSA-N
Synonym 4-pentyl-1,1':4',1-terphenyl-4-carbonitrile, 4-cyano-4-n-pentyl-p-terphenyl, 4-cyano-4-pentyl-p-terphenyl, 1,1':4',1-terphenyl-4-carbonitrile, 4-pentyl, 4-4-4-pentylphenyl phenyl benzonitrile, pubchem9116, 4-pentyl-p-terphenyl-4-carbonitrile, 4-cyano-4'-pentyl-p-terphenyl
PubChem CID 104707
IUPAC Name 4-[4-(4-pentylphenyl)phenyl]benzonitrile
SMILES CCCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)C#N

Specifications

Melting Point 128°C to 130°C
Quantity 0.5 g
UN Number UN3439
Formula Weight 325.45
Percent Purity 99%
Chemical Name or Material 4-Cyano-4''-n-pentyl-p-terphenyl
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 259-028-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only