missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4'-n-octyloxybiphenyl 98.0+%, TCI America™

Catalog No. p-7125561
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26185G 5 g
C261825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26185G Supplier TCI America Supplier No. C26185G
Only null left

Chemical Identifiers

CAS 52364-73-5
Molecular Formula C21H25NO
Molecular Weight (g/mol) 307.437
MDL Number MFCD00075145
InChI Key GPGGNNIMKOVSAG-UHFFFAOYSA-N
Synonym 4'-octyloxy-4-biphenylcarbonitrile, 4'-octyloxy-1,1'-biphenyl-4-carbonitrile, 4-cyano-4'-octyloxybiphenyl, 8ocb, octyloxycyanobiphenyl, 4-cyano-4'-octoxybiphenyl, 1,1'-biphenyl-4-carbonitrile, 4'-octyloxy, m 24 liquid crystal, 4-octyloxycyanodiphenyl, 4-4-octyloxyphenyl benzonitrile
PubChem CID 104173
IUPAC Name 4-(4-octoxyphenyl)benzonitrile
SMILES CCCCCCCCOC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 54°C
Color White
Quantity 5 g
Formula Weight 307.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-4′-n-octyloxybiphenyl
RTECSNumber DV1764500
TSCA Yes