Learn More
4-Cyano-4'-n-hexylbiphenyl, 97%, Thermo Scientific™
$173.71 - $173.71
Chemical Identifiers
| CAS | 41122-70-7 |
|---|---|
| Molecular Formula | C19H21N |
| Molecular Weight (g/mol) | 263.384 |
| MDL Number | MFCD00075144 |
| InChI Key | VADSDVGLFDVIMG-UHFFFAOYSA-N |
| Synonym | 4-hexyl-4'-cyanobiphenyl, 4-cyano-4'-hexylbiphenyl, 4'-hexyl-4-biphenylcarbonitrile, 4-n-hexyl-4'-cyanobiphenyl, 4'-hexyl-1,1'-biphenyl-4-carbonitrile, 4'-hexylbiphenyl-4-carbonitrile, ccris 6044, 1,1'-biphenyl-4-carbonitrile, 4'-hexyl, 4-4-hexylphenyl benzonitrile |
| PubChem CID | 38764 |
| IUPAC Name | 4-(4-hexylphenyl)benzonitrile |
| SMILES | CCCCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5582503
|
Thermo Scientific Chemicals
H5582503 |
1 g |
Each for $173.71
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications4-Cyano-4′-n-hexylbiphenyl, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a liquid crystal monomer.
Solubility
Sparingly Soluble in water (3.2E-5 g/L) (25°C).
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
| 41122-70-7 | |
| 263.384 | |
| VADSDVGLFDVIMG-UHFFFAOYSA-N | |
| 38764 | |
| CCCCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C#N |
| C19H21N | |
| MFCD00075144 | |
| 4-hexyl-4'-cyanobiphenyl, 4-cyano-4'-hexylbiphenyl, 4'-hexyl-4-biphenylcarbonitrile, 4-n-hexyl-4'-cyanobiphenyl, 4'-hexyl-1,1'-biphenyl-4-carbonitrile, 4'-hexylbiphenyl-4-carbonitrile, ccris 6044, 1,1'-biphenyl-4-carbonitrile, 4'-hexyl, 4-4-hexylphenyl benzonitrile | |
| 4-(4-hexylphenyl)benzonitrile |
Specifications
| 41122-70-7 | |
| C19H21N | |
| 1 g | |
| Sparingly Soluble in water (3.2E-5g/L) (25°C). | |
| CCCCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C#N | |
| 263.384 | |
| 263.38 | |
| 4-Cyano-4'-n-hexylbiphenyl |
| 113°C (235°F) | |
| MFCD00075144 | |
| 4-hexyl-4'-cyanobiphenyl, 4-cyano-4'-hexylbiphenyl, 4'-hexyl-4-biphenylcarbonitrile, 4-n-hexyl-4'-cyanobiphenyl, 4'-hexyl-1,1'-biphenyl-4-carbonitrile, 4'-hexylbiphenyl-4-carbonitrile, ccris 6044, 1,1'-biphenyl-4-carbonitrile, 4'-hexyl, 4-4-hexylphenyl benzonitrile | |
| VADSDVGLFDVIMG-UHFFFAOYSA-N | |
| 4-(4-hexylphenyl)benzonitrile | |
| 38764 | |
| 97% |
Safety and Handling
GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 255-228-5
TSCA : Yes
Recommended Storage : Ambient temperatures