missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4'-hydroxybiphenyl 98.0+%, TCI America™

Catalog No. p-7130582
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05235G 5 g
H052325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05235G Supplier TCI America Supplier No. H05235G

Chemical Identifiers

CAS 19812-93-2
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.221
MDL Number MFCD00059625
InChI Key ZRMIETZFPZGBEB-UHFFFAOYSA-N
Synonym 4-cyano-4'-hydroxybiphenyl, 4'-hydroxy-4-biphenylcarbonitrile, 4-4-hydroxyphenyl benzonitrile, 4'-hydroxybiphenyl-4-carbonitrile, 4'-cyano-4-hydroxybiphenyl, 4'-hydroxy-1,1'-biphenyl-4-carbonitrile, 4'-cyano-4-biphenylol, 4-cyano-4'-hydroxy-biphenyl, 4'-hydroxy-4-cyanobiphenyl, 4-hydroxy-4-biphenylcarbonitrile
PubChem CID 140610
IUPAC Name 4-(4-hydroxyphenyl)benzonitrile
SMILES C1=CC(=CC=C1C#N)C2=CC=C(C=C2)O

Specifications

Melting Point 198°C
Color White-Yellow
Quantity 5 g
Formula Weight 195.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-4′-hydroxybiphenyl
EINECSNumber (4)-1822
TSCA No