missing translation for 'onlineSavingsMsg'
Learn More

4'-Cyano-4-hexyloxybiphenyl 98.0+%, TCI America™

Catalog No. C31555G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31555G 5 g
C315525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31555G Supplier TCI America Supplier No. C31555G
Only null left

Chemical Identifiers

CAS 41424-11-7
Molecular Formula C19H21NO
Molecular Weight (g/mol) 279.38
MDL Number MFCD00134145
InChI Key YUYXUPYNSOFWFV-UHFFFAOYSA-N
Synonym 4'-hexyloxy-4-biphenylcarbonitrile, 6ocb, liquid crystal 549, 1,1'-biphenyl-4-carbonitrile, 4'-hexyloxy, 4-hexyloxycyanodiphenyl, 4-cyano-4'-hexoxybiphenyl, 4-4-hexyloxyphenyl benzonitrile, 4'-hexyloxy-1,1'-biphenyl-4-carbonitrile
PubChem CID 162465
IUPAC Name 4'-(hexyloxy)-[1,1'-biphenyl]-4-carbonitrile
SMILES CCCCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C#N

Specifications

Melting Point 58°C
Color White
Quantity 5 g
Formula Weight 279.38
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Cyano-4-hexyloxybiphenyl
RTECSNumber DV1764200
TSCA Yes