missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4'-hexylbiphenyl 98.0+%, TCI America™

Catalog No. C31545G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31545G 5 g
C31541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31545G Supplier TCI America Supplier No. C31545G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41122-70-7
Molecular Formula C19H21N
Molecular Weight (g/mol) 263.384
MDL Number MFCD00075144
InChI Key VADSDVGLFDVIMG-UHFFFAOYSA-N
Synonym 4-hexyl-4'-cyanobiphenyl, 4-cyano-4'-hexylbiphenyl, 4'-hexyl-4-biphenylcarbonitrile, 4-n-hexyl-4'-cyanobiphenyl, 4'-hexyl-1,1'-biphenyl-4-carbonitrile, 4'-hexylbiphenyl-4-carbonitrile, ccris 6044, 1,1'-biphenyl-4-carbonitrile, 4'-hexyl, 4-4-hexylphenyl benzonitrile
PubChem CID 38764
IUPAC Name 4-(4-hexylphenyl)benzonitrile
SMILES CCCCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 14°C
Color Yellow
Boiling Point 185°C
Quantity 5 g
UN Number 3082
Formula Weight 263.38
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Cyano-4′-hexylbiphenyl
EINECSNumber (4)-0831
TSCA Yes