missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4'-heptylbiphenyl 95.0+%, TCI America™

Catalog No. H08121G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08121G 1 g
H08125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08121G Supplier TCI America Supplier No. H08121G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41122-71-8
Molecular Formula C20H23N
Molecular Weight (g/mol) 277.411
MDL Number MFCD00074915
InChI Key ZGOWXOZNUNZPAV-UHFFFAOYSA-N
Synonym 4-cyano-4'-heptylbiphenyl, 4'-heptyl-1,1'-biphenyl-4-carbonitrile, 4'-heptyl-4-biphenylcarbonitrile, 1,1'-biphenyl-4-carbonitrile, 4'-heptyl, 4-cyano-4'-n-heptylbiphenyl, 4-4-heptylphenyl benzonitrile, 4-heptyl-4'-cyanobiphenyl, 4-cyano-4'-n-heptyl-biphenyl, 4'-heptylbiphenyl-4-carbonitrile, 4-4-heptylphenyl benzenecarbonitrile
PubChem CID 170468
IUPAC Name 4-(4-heptylphenyl)benzonitrile
SMILES CCCCCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 36°C
Color White
Quantity 1 g
Formula Weight 277.41
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Cyano-4′-heptylbiphenyl