missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4'-ethoxybiphenyl 98.0+%, TCI America™

Catalog No. C31535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31535G 5 g
C315325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31535G Supplier TCI America Supplier No. C31535G
Only null left

Chemical Identifiers

CAS 58743-78-5
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.275
MDL Number MFCD01218033
InChI Key VETJRGXWDLHERN-UHFFFAOYSA-N
Synonym 4'-ethoxy-1,1'-biphenyl-4-carbonitrile, 4-cyano-4'-ethoxybiphenyl, 4-ethoxy-4'-cyanobiphenyl, 4-4-ethoxyphenyl benzonitrile, 4-ethoxy-1,1'-biphenyl-4'-carbonitrile, 1,1'-biphenyl-4-carbonitrile, 4'-ethoxy, 4'-ethoxy-4-cyanobiphenyl, 4-4-ethoxyphenyl benzenecarbonitrile, pubchem2524
PubChem CID 93893
IUPAC Name 4-(4-ethoxyphenyl)benzonitrile
SMILES CCOC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 102°C
Color White
Quantity 5 g
Formula Weight 223.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-4′-ethoxybiphenyl