missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Cyano-4-decyloxybiphenyl 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Synonym | 4′-Decyloxy-4-biphenylcarbonitrile, 4′-(Decyloxy)-[1,1′-biphenyl]-4-carbonitrile, 10OCB |
| Formula Weight | 335.49 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4′-Cyano-4-decyloxybiphenyl |
Safety and Handling
| TSCA | Yes |