missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyano-3-tetrahydrothiophenone 98.0+%, TCI America™
Supplier: TCI America C116425G
Specifications
| 4-Cyano-3-tetrahydrothiophenone | |
| 71°C | |
| 125°C | |
| MFCD00059049 | |
| 4-Cyano-3-oxotetrahydrothiophene | |
| C1C(C(=O)CS1)C#N | |
| 127.161 | |
| 127.16 | |
| Crystalline Powder |
| 16563-14-7 | |
| White-Yellow | |
| C5H5NOS | |
| 25 g | |
| OHPSBDAUCJNDHP-UHFFFAOYSA-N | |
| 4-oxothiolane-3-carbonitrile | |
| 326676 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 16563-14-7 | |
| 127.161 | |
| OHPSBDAUCJNDHP-UHFFFAOYSA-N | |
| 326676 | |
| C1C(C(=O)CS1)C#N |
| C5H5NOS | |
| MFCD00059049 | |
| 4-Cyano-3-oxotetrahydrothiophene | |
| 4-oxothiolane-3-carbonitrile |
Safety and Handling
TSCA : No