missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-3-tetrahydrothiophenone 98.0+%, TCI America™

Catalog No. C11645G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11645G 5 g
C116425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C11645G Supplier TCI America Supplier No. C11645G
Only null left

Chemical Identifiers

CAS 16563-14-7
Molecular Formula C5H5NOS
Molecular Weight (g/mol) 127.161
MDL Number MFCD00059049
InChI Key OHPSBDAUCJNDHP-UHFFFAOYSA-N
Synonym 4-Cyano-3-oxotetrahydrothiophene
PubChem CID 326676
IUPAC Name 4-oxothiolane-3-carbonitrile
SMILES C1C(C(=O)CS1)C#N

Specifications

Melting Point 71°C
Color White-Yellow
Boiling Point 125°C
Quantity 5 g
Formula Weight 127.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-3-tetrahydrothiophenone