missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-2-fluoropyridine 99.0+%, TCI America™

$119.30 - $359.76

Chemical Identifiers

CAS 3939-14-8
Molecular Formula C6H3FN2
Molecular Weight (g/mol) 122.102
MDL Number MFCD09027299
InChI Key DYQMGPSBUBTNJZ-UHFFFAOYSA-N
PubChem CID 12002587
IUPAC Name 2-fluoropyridine-4-carbonitrile
SMILES C1=CN=C(C=C1C#N)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C33835G
View Documents
TCI America
C33835G
5 g
Each for $119.30
Only null left
 
C338325G
View Documents
TCI America
C338325G
25 g
Each for $359.76
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3939-14-8
122.102
DYQMGPSBUBTNJZ-UHFFFAOYSA-N
2-fluoropyridine-4-carbonitrile
C6H3FN2
MFCD09027299
12002587
C1=CN=C(C=C1C#N)F

Specifications

3939-14-8
White
MFCD09027299
3439
DYQMGPSBUBTNJZ-UHFFFAOYSA-N
2-fluoropyridine-4-carbonitrile
12002587
≥99.0% (GC)
4-Cyano-2-fluoropyridine
34°C
C6H3FN2
5 g
2-fluoroisonicotinonitrile, 4-cyano-2-fluoropyridine, 2-fluoro-4-cyanopyridine, 4-pyridinecarbonitrile, 2-fluoro, 2-fluoro-4-pyridinecarbonitrile, 2-fluoro-isonicotinonitrile, 4-pyridinecarbonitrile,2-fluoro, acmc-1bnd8, 4-cyano-2-fluoro pyridine, ksc581g3h
C1=CN=C(C=C1C#N)F
122.102
122.10
Crystalline Lumps
Videos
SDS
Documents

Documents