missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyano-2-fluoropyridine 99.0+%, TCI America™
Supplier: TCI America C33835G
Specifications
| 4-Cyano-2-fluoropyridine | |
| 34°C | |
| C6H3FN2 | |
| 5 g | |
| 2-fluoroisonicotinonitrile, 4-cyano-2-fluoropyridine, 2-fluoro-4-cyanopyridine, 4-pyridinecarbonitrile, 2-fluoro, 2-fluoro-4-pyridinecarbonitrile, 2-fluoro-isonicotinonitrile, 4-pyridinecarbonitrile,2-fluoro, acmc-1bnd8, 4-cyano-2-fluoro pyridine, ksc581g3h | |
| C1=CN=C(C=C1C#N)F | |
| 122.102 | |
| 122.10 | |
| Crystalline Lumps |
| 3939-14-8 | |
| White | |
| MFCD09027299 | |
| 3439 | |
| DYQMGPSBUBTNJZ-UHFFFAOYSA-N | |
| 2-fluoropyridine-4-carbonitrile | |
| 12002587 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 3939-14-8 | |
| 122.102 | |
| DYQMGPSBUBTNJZ-UHFFFAOYSA-N | |
| 2-fluoropyridine-4-carbonitrile |
| C6H3FN2 | |
| MFCD09027299 | |
| 12002587 | |
| C1=CN=C(C=C1C#N)F |