Learn More
4-Cyano-1-butyne, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 455590050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 4-Cyano-1-butyne | |
| 0.9063g/mL | |
| 65.5°C | |
| 96% min. (GC) | |
| NCCH2CH2CCH | |
| 0.9063 | |
| VMUWIDHKAIGONP-UHFFFAOYSA-N | |
| pent-4-ynenitrile | |
| 140560 | |
| 97% |
| 19596-07-7 | |
| 79.0°C (39.0 mmHg) | |
| Authentic | |
| C5H5N | |
| 5g | |
| 4-cyano-1-butyne, 4-pentynenitrile, 1-cyano-3-butyne | |
| C#CCCC#N | |
| 79.1 | |
| 79.1 |
Chemical Identifiers
| 19596-07-7 | |
| 79.1 | |
| 4-cyano-1-butyne, 4-pentynenitrile, 1-cyano-3-butyne | |
| pent-4-ynenitrile |
| C5H5N | |
| VMUWIDHKAIGONP-UHFFFAOYSA-N | |
| 140560 | |
| C#CCCC#N |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy
GHS Signal Word: Danger
RUO â Research Use Only