missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-1-butyne, 97%

Catalog No. p-7077565
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455590010 1 g
AC455590050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC455590010 Supplier Thermo Scientific Chemicals Supplier No. 455590010
Only null left

CAS: 19596-07-7 | C5H5N | 79.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19596-07-7
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.1
InChI Key VMUWIDHKAIGONP-UHFFFAOYSA-N
Synonym 4-cyano-1-butyne, 4-pentynenitrile, 1-cyano-3-butyne
PubChem CID 140560
IUPAC Name pent-4-ynenitrile
SMILES C#CCCC#N

Specifications

Density 0.9063g/mL
Boiling Point 79.0°C (39.0 mmHg)
Flash Point 65.5°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula NCCH2CH2CCH
Quantity 1 g
Specific Gravity 0.9063
Formula Weight 79.1
Percent Purity 97%
Chemical Name or Material 4-Cyano-1-butyne
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy

RUO – Research Use Only