missing translation for 'onlineSavingsMsg'
Learn More

4-Cumylphenol, 97%

Catalog No. AAB2341030
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2341030 250 g
AAB2341022 100 g
AAB234100B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2341030 Supplier Thermo Scientific Chemicals Supplier No. B2341030
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 599-64-4 | C15H16O | 212.292 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 599-64-4
Molecular Formula C15H16O
Molecular Weight (g/mol) 212.292
MDL Number MFCD00002365
InChI Key QBDSZLJBMIMQRS-UHFFFAOYSA-N
Synonym 4-cumylphenol, p-cumylphenol, 4-2-phenylpropan-2-yl phenol, p-cumyl phenol, phenol, 4-1-methyl-1-phenylethyl, 4-1-methyl-1-phenylethyl phenol, 4-alpha-cumylphenol, 4-dimethylphenylmethyl phenol, 4-hydroxydiphenyldimethylmethane, 2-phenyl-2-4-hydroxyphenyl propane
PubChem CID 11742
ChEBI CHEBI:35092
IUPAC Name 4-(2-phenylpropan-2-yl)phenol
SMILES CC(C)(C1=CC=CC=C1)C2=CC=C(C=C2)O

Specifications

Melting Point 70°C to 73°C
Density 1.115
Boiling Point 189°C to 191°C (10 mmHg)
Flash Point 160°C (320°F)
Quantity 250 g
Beilstein 1870517
Formula Weight 212.29
Percent Purity 97%
Chemical Name or Material 4-Cumylphenol
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 209-968-0
RTECSNumber SL1942450
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only