missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorothiophenol, 97%

Catalog No. AAB2383814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2383814 25 g
AAB2383822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2383814 Supplier Thermo Scientific Chemicals Supplier No. B2383814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 106-54-7 | C6H5ClS | 144.616 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106-54-7
Molecular Formula C6H5ClS
Molecular Weight (g/mol) 144.616
MDL Number MFCD00004847
InChI Key VZXOZSQDJJNBRC-UHFFFAOYSA-N
Synonym 4-chlorothiophenol, p-chlorothiophenol, benzenethiol, 4-chloro, p-chlorobenzenethiol, p-chlorthiofenol, p-chlorophenylmercaptan, benzenethiol, p-chloro, p-mercaptochlorobenzene, 4-chlorophenylmercaptan, 1-chloro-4-mercaptobenzene
PubChem CID 7815
IUPAC Name 4-chlorobenzenethiol
SMILES C1=CC(=CC=C1S)Cl

Specifications

Melting Point 48°C to 52°C
Boiling Point 205°C to 207°C
Flash Point 81°C (177°F)
Quantity 25 g
UN Number UN1759
Beilstein 605971
Sensitivity Air Sensitive
Formula Weight 144.62
Percent Purity 97%
Chemical Name or Material 4-Chlorothiophenol
Hazard Category H302-H314
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber 203-408-9
RTECSNumber DC1050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only