missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorothioanisole 98.0+%, TCI America™

Catalog No. C115610G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C115610G 10 g
C115625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. C115610G missing translation for 'mfr' TCI America missing translation for 'supplierNo' C115610G

Chemical Identifiers

CAS 123-09-1
Molecular Formula C7H7ClS
Molecular Weight (g/mol) 158.643
MDL Number MFCD00013643
InChI Key KIQQUVJOLVCZKG-UHFFFAOYSA-N
Synonym 4-chlorothioanisole, 4-chlorophenyl methyl sulfide, p-chlorothioanisole, 1-chloro-4-methylthio benzene, p-chlorophenyl methyl sulfide, methyl 4-chlorophenyl sulfide, benzene, 1-chloro-4-methylthio, p-chlophenyl methyl sulfide, 1-chloro-4-methylsulfanyl benzene, 4-chloro thioanisole
PubChem CID 31243
IUPAC Name 1-chloro-4-methylsulfanylbenzene
SMILES CSC1=CC=C(C=C1)Cl

Specifications

Color Yellow
Boiling Point 224°C
Quantity 10 g
Formula Weight 158.64
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chlorothioanisole
RTECSNumber WQ3840000
TSCA Yes