missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorothioanisole, 98%

Catalog No. p-7081175
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1102506 5 g
AAA1102514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1102506 Supplier Thermo Scientific Chemicals Supplier No. A1102506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 123-09-1 | C7H7ClS | 158.643 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 123-09-1
Molecular Formula C7H7ClS
Molecular Weight (g/mol) 158.643
MDL Number MFCD00013643
InChI Key KIQQUVJOLVCZKG-UHFFFAOYSA-N
Synonym 4-chlorothioanisole, 4-chlorophenyl methyl sulfide, p-chlorothioanisole, 1-chloro-4-methylthio benzene, p-chlorophenyl methyl sulfide, methyl 4-chlorophenyl sulfide, benzene, 1-chloro-4-methylthio, p-chlophenyl methyl sulfide, 1-chloro-4-methylsulfanyl benzene, 4-chloro thioanisole
PubChem CID 31243
IUPAC Name 1-chloro-4-methylsulfanylbenzene
SMILES CSC1=CC=C(C=C1)Cl

Specifications

Density 1.22
Boiling Point 223°C
Flash Point 90°C (194°F)
Odor Unpleasant
Refractive Index 1.6
Quantity 5 g
Beilstein 2041509
Formula Weight 158.65
Percent Purity 98%
Chemical Name or Material 4-Chlorothioanisole
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 204-600-5
RTECSNumber WQ3840000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only