missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorostyrene (stabilized with TBC) 98.0+%, TCI America™

Catalog No. C029010ML
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C029010ML 10 mL
C029025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C029010ML Supplier TCI America Supplier No. C029010ML

Chemical Identifiers

CAS 1073-67-2
Molecular Formula C8H7Cl
Molecular Weight (g/mol) 138.59
MDL Number MFCD00000632
InChI Key KTZVZZJJVJQZHV-UHFFFAOYSA-N
Synonym 4-chlorostyrene, p-chlorostyrene, 1-chloro-4-vinylbenzene, parachlorostyrene, benzene, 1-chloro-4-ethenyl, styrene, p-chloro, styrene, 4-chloro, unii-t0j05u220f, para-chlorostyrene
PubChem CID 14085
IUPAC Name 1-chloro-4-ethenylbenzene
SMILES ClC1=CC=C(C=C)C=C1

Specifications

Color Yellow
Quantity 10 mL
Formula Weight 138.59
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chlorostyrene (stabilized with TBC)
EINECSNumber (3)-3443
RTECSNumber CZ0533000
TSCA Yes
Recommended Storage Refrigerator