Learn More
4-Chlorostyrene, 99%, stabilized
CAS: 1073-67-2 | C8H7Cl | 138.59 g/mol
$169.04 - $169.04
Chemical Identifiers
| CAS | 1073-67-2 |
|---|---|
| Molecular Formula | C8H7Cl |
| Molecular Weight (g/mol) | 138.59 |
| MDL Number | MFCD00000632 |
| InChI Key | KTZVZZJJVJQZHV-UHFFFAOYSA-N |
| Synonym | 4-chlorostyrene, p-chlorostyrene, 1-chloro-4-vinylbenzene, parachlorostyrene, benzene, 1-chloro-4-ethenyl, styrene, p-chloro, styrene, 4-chloro, unii-t0j05u220f, para-chlorostyrene |
| PubChem CID | 14085 |
| IUPAC Name | 1-chloro-4-ethenylbenzene |
| SMILES | ClC1=CC=C(C=C)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC110090100
|
Thermo Scientific Chemicals
110090100 |
10 g | Glass bottle |
Each for $169.04
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1073-67-2 | |
| 138.59 | |
| KTZVZZJJVJQZHV-UHFFFAOYSA-N | |
| 14085 | |
| ClC1=CC=C(C=C)C=C1 |
| C8H7Cl | |
| MFCD00000632 | |
| 4-chlorostyrene, p-chlorostyrene, 1-chloro-4-vinylbenzene, parachlorostyrene, benzene, 1-chloro-4-ethenyl, styrene, p-chloro, styrene, 4-chloro, unii-t0j05u220f, para-chlorostyrene | |
| 1-chloro-4-ethenylbenzene |
Specifications
| 1073-67-2 | |
| 100.0 | |
| Colorless to Yellow | |
| 192°C | |
| Authentic | |
| Glass bottle | |
| 1.5645 to 1.5665 | |
| MFCD00000632 | |
| 05, II, 367 | |
| 4-chlorostyrene, p-chlorostyrene, 1-chloro-4-vinylbenzene, parachlorostyrene, benzene, 1-chloro-4-ethenyl, styrene, p-chloro, styrene, 4-chloro, unii-t0j05u220f, para-chlorostyrene | |
| ClC1=CC=C(C=C)C=C1 | |
| 138.59 | |
| 14085 | |
| 99% | |
| 4-Chlorostyrene, 99% |
| 98.5 | |
| -16°C | |
| 1.1000g/mL | |
| 60°C | |
| 98.5% min. (GC) | |
| C8H7Cl | |
| H2C=CHC6H4Cl | |
| 10 g | |
| 1.1 | |
| KTZVZZJJVJQZHV-UHFFFAOYSA-N | |
| 1-chloro-4-ethenylbenzene | |
| 0.1% tert.-butylcatechol | |
| 138.6 | |
| Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
May produce an allergic reaction.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Warning
EINECSNumber : 214-028-8
TSCA : TSCA
RUO – Research Use Only