missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorostyrene, 98+%, stab

Catalog No. AAB2051218
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2051218 50 g
AAB2051209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2051218 Supplier Thermo Scientific Chemicals Supplier No. B2051218

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1073-67-2 | C8H7Cl | 138.59 g/mol

4-Chlorostyrene is used in the preparation of new bis(pyrazolyl)borato olefin complexes of copper(I). It is also employed in the investigation of regioselectivity in the cationic Heck reaction of 4-substituted styrenes. Further, it plays an important role in the preparation of 4-[(4-chloro-phenyl)-thioacetyl]-morpholine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorostyrene is used in the preparation of new bis(pyrazolyl)borato olefin complexes of copper(I). It is also employed in the investigation of regioselectivity in the cationic Heck reaction of 4-substituted styrenes. Further, it plays an important role in the preparation of 4-[(4-chloro-phenyl)-thioacetyl]-morpholine.

Solubility
Immiscible with water.

Notes
Incompatible with acids, bases, oxidizing agents, halogens, metal salts and polymerizing initiators. Store in a cool place.

Chemical Identifiers

CAS 1073-67-2
Molecular Formula C8H7Cl
Molecular Weight (g/mol) 138.59
MDL Number MFCD00000632
InChI Key KTZVZZJJVJQZHV-UHFFFAOYSA-N
Synonym 4-chlorostyrene, p-chlorostyrene, 1-chloro-4-vinylbenzene, parachlorostyrene, benzene, 1-chloro-4-ethenyl, styrene, p-chloro, styrene, 4-chloro, unii-t0j05u220f, para-chlorostyrene
PubChem CID 14085
IUPAC Name 1-chloro-4-ethenylbenzene
SMILES ClC1=CC=C(C=C)C=C1

Specifications

Melting Point -16°C
Density 1.089
Boiling Point 191°C to 192°C
Flash Point 67°C (153°F)
Odor Characteristic
Refractive Index 1.565
Quantity 50 g
Beilstein 1098859
Solubility Information Immiscible with water.
Formula Weight 138.6
Percent Purity 99%
Chemical Name or Material 4-Chlorostyrene, stabilized
Show More Show Less
Hazard Category H227-H302-H315-H319
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 214-028-8
RTECSNumber CZ0533000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only