missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorosalicylic Acid 98.0+%, TCI America™
$126.76 - $1168.76
Chemical Identifiers
| CAS | 5106-98-9 |
|---|---|
| Molecular Formula | C7H5ClO3 |
| Molecular Weight (g/mol) | 172.56 |
| MDL Number | MFCD00002449 |
| InChI Key | LWXFCZXRFBUOOR-UHFFFAOYSA-N |
| Synonym | 4-chlorosalicylic acid, 4-chloro salicylic acid, benzoic acid, 4-chloro-2-hydroxy, 2-hydroxy-4-chlorobenzoic acid, 4-chloro-2-hydroxy benzoic acid, 4-chloro-2-hydroxy-benzoic acid, salicylic acid, 4-chloro, 4-chlorosalicylicacid, pubchem3845, 4-chloro-salicylic acid |
| PubChem CID | 78782 |
| IUPAC Name | 4-chloro-2-hydroxybenzoic acid |
| SMILES | OC(=O)C1=CC=C(Cl)C=C1O |
Chemical Identifiers
| 5106-98-9 | |
| 172.56 | |
| LWXFCZXRFBUOOR-UHFFFAOYSA-N | |
| 78782 | |
| OC(=O)C1=CC=C(Cl)C=C1O |
| C7H5ClO3 | |
| MFCD00002449 | |
| 4-chlorosalicylic acid, 4-chloro salicylic acid, benzoic acid, 4-chloro-2-hydroxy, 2-hydroxy-4-chlorobenzoic acid, 4-chloro-2-hydroxy benzoic acid, 4-chloro-2-hydroxy-benzoic acid, salicylic acid, 4-chloro, 4-chlorosalicylicacid, pubchem3845, 4-chloro-salicylic acid | |
| 4-chloro-2-hydroxybenzoic acid |
Specifications
| 5106-98-9 | |
| C7H5ClO3 | |
| 25 g | |
| LWXFCZXRFBUOOR-UHFFFAOYSA-N | |
| 4-chloro-2-hydroxybenzoic acid | |
| 78782 | |
| ≥98.0% (GC,T) | |
| 4-Chlorosalicylic Acid |
| 217°C | |
| MFCD00002449 | |
| 4-chlorosalicylic acid, 4-chloro salicylic acid, benzoic acid, 4-chloro-2-hydroxy, 2-hydroxy-4-chlorobenzoic acid, 4-chloro-2-hydroxy benzoic acid, 4-chloro-2-hydroxy-benzoic acid, salicylic acid, 4-chloro, 4-chlorosalicylicacid, pubchem3845, 4-chloro-salicylic acid | |
| OC(=O)C1=CC=C(Cl)C=C1O | |
| 172.56 | |
| 172.56 | |
| Crystalline Powder |
Safety and Handling
TSCA : No