missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorosalicylic Acid 98.0+%, TCI America™

Catalog No. C1282500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1282500G 500 g
C128225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1282500G Supplier TCI America Supplier No. C1282500G

Chemical Identifiers

CAS 5106-98-9
Molecular Formula C7H5ClO3
Molecular Weight (g/mol) 172.56
MDL Number MFCD00002449
InChI Key LWXFCZXRFBUOOR-UHFFFAOYSA-N
Synonym 4-chlorosalicylic acid, 4-chloro salicylic acid, benzoic acid, 4-chloro-2-hydroxy, 2-hydroxy-4-chlorobenzoic acid, 4-chloro-2-hydroxy benzoic acid, 4-chloro-2-hydroxy-benzoic acid, salicylic acid, 4-chloro, 4-chlorosalicylicacid, pubchem3845, 4-chloro-salicylic acid
PubChem CID 78782
IUPAC Name 4-chloro-2-hydroxybenzoic acid
SMILES OC(=O)C1=CC=C(Cl)C=C1O

Specifications

Melting Point 217°C
Quantity 500 g
Formula Weight 172.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorosalicylic Acid