missing translation for 'onlineSavingsMsg'
Learn More

4-Chlororesorcinol 98.0+%, TCI America™

Catalog No. C0285500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0285500G 500 g
C028525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0285500G Supplier TCI America Supplier No. C0285500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95-88-5
Molecular Formula C6H5ClO2
Molecular Weight (g/mol) 144.55
MDL Number MFCD00002273
InChI Key JQVAPEJNIZULEK-UHFFFAOYSA-N
Synonym 4-chlororesorcinol, 1,3-benzenediol, 4-chloro, p-chlororesorcinol, 4-chlororesorcin, resorcinol, 4-chloro, 6-chlororesorcinol, 4-chloro-1,3-dihydroxybenzene, 4-chloro-1,3-benzenediol, 2,4-dihydroxychlorobenzene, 1,3-dihydroxy-4-chlorobenzene
PubChem CID 1731
IUPAC Name 4-chlorobenzene-1,3-diol
SMILES OC1=CC=C(Cl)C(O)=C1

Specifications

Melting Point 107°C
Boiling Point 256°C
Quantity 500 g
Formula Weight 144.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlororesorcinol
EINECSNumber (3)-0901
RTECSNumber VH0450000
TSCA Yes