missing translation for 'onlineSavingsMsg'
Learn More

4-Chlororesorcinol, 98%

CAS: 95-88-5 | C6H5ClO2 | 144.55 g/mol

$89.15 - $89.15

Chemical Identifiers

CAS 95-88-5
Molecular Formula C6H5ClO2
Molecular Weight (g/mol) 144.55
MDL Number MFCD00002273
InChI Key JQVAPEJNIZULEK-UHFFFAOYSA-N
Synonym 4-chlororesorcinol, 1,3-benzenediol, 4-chloro, p-chlororesorcinol, 4-chlororesorcin, resorcinol, 4-chloro, 6-chlororesorcinol, 4-chloro-1,3-dihydroxybenzene, 4-chloro-1,3-benzenediol, 2,4-dihydroxychlorobenzene, 1,3-dihydroxy-4-chlorobenzene
PubChem CID 1731
IUPAC Name 4-chlorobenzene-1,3-diol
SMILES OC1=CC=C(Cl)C(O)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC110061000
SDS View Documents
Thermo Scientific Chemicals
110061000
100 g Glass bottle
Each for $89.15
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

95-88-5
144.55
JQVAPEJNIZULEK-UHFFFAOYSA-N
1731
OC1=CC=C(Cl)C(O)=C1
C6H5ClO2
MFCD00002273
4-chlororesorcinol, 1,3-benzenediol, 4-chloro, p-chlororesorcinol, 4-chlororesorcin, resorcinol, 4-chloro, 6-chlororesorcinol, 4-chloro-1,3-dihydroxybenzene, 4-chloro-1,3-benzenediol, 2,4-dihydroxychlorobenzene, 1,3-dihydroxy-4-chlorobenzene
4-chlorobenzene-1,3-diol

Specifications

95-88-5
100.0
Beige
148°C
98%
C6H5ClO2
100 g
4-chlororesorcinol, 1,3-benzenediol, 4-chloro, p-chlororesorcinol, 4-chlororesorcin, resorcinol, 4-chloro, 6-chlororesorcinol, 4-chloro-1,3-dihydroxybenzene, 4-chloro-1,3-benzenediol, 2,4-dihydroxychlorobenzene, 1,3-dihydroxy-4-chlorobenzene
JQVAPEJNIZULEK-UHFFFAOYSA-N
4-chlorobenzene-1,3-diol
1731
98%
4-Chlororesorcinol, 98%
97.5
106°C to 110°C
147°C (18.0 mmHg)
Authentic
Glass bottle
MFCD00002273
06, II, 818
Solubility in water: 1000g/L (25°C)
OC1=CC=C(Cl)C(O)=C1
144.55
144.56
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove

GHS Signal Word: Warning

EINECSNumber : 202-462-0

RTECSNumber : VH0450000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only