missing translation for 'onlineSavingsMsg'
Learn More

4-Chloropyrazole 98.0+%, TCI America™

Catalog No. C28885G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28885G 5 g
C288825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28885G Supplier TCI America Supplier No. C28885G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15878-00-9
Molecular Formula C3H3ClN2
Molecular Weight (g/mol) 102.521
MDL Number MFCD00159634
InChI Key BADSZRMNXWLUKO-UHFFFAOYSA-N
Synonym 4-chloropyrazole, 1h-pyrazole, 4-chloro, 4-chloro pyrazole, 1h-pyrazole, 4-chloro-9ci, pyrazole, 4-chloro, zlchem 159, pubchem14005, 5-23-04-00164 beilstein handbook reference
PubChem CID 27524
IUPAC Name 4-chloro-1H-pyrazole
SMILES C1=C(C=NN1)Cl

Specifications

Melting Point 77°C
Color White
Boiling Point 220°C
Quantity 5 g
Formula Weight 102.52
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloropyrazole
RTECSNumber UQ6445000
TSCA No