accessibility menu, dialog, popup

UserName

ClNHN

CAS RN 15878-00-9

IUPAC Name: 4-chloro-1H-pyrazole
Synonyms: 1H-pyrazole, 4-chloro- (9CI)
Molecular Weight (g/mol): 102.52
Molecular Formula: C3H3ClN2
InChi Key: BADSZRMNXWLUKO-UHFFFAOYSA-N
SMILES: ClC1=CNN=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)