missing translation for 'onlineSavingsMsg'
Learn More

4-Chloropyrazole, 98%

Catalog No. AC459920050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459920050 5 g
AC459920250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459920050 Supplier Thermo Scientific Chemicals Supplier No. 459920050

CAS: 15878-00-9 | C3H3ClN2 | 102.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15878-00-9
Molecular Formula C3H3ClN2
Molecular Weight (g/mol) 102.52
InChI Key BADSZRMNXWLUKO-UHFFFAOYSA-N
Synonym 4-chloropyrazole, 1h-pyrazole, 4-chloro, 4-chloro pyrazole, 1h-pyrazole, 4-chloro-9ci, pyrazole, 4-chloro, zlchem 159, pubchem14005, 5-23-04-00164 beilstein handbook reference
PubChem CID 27524
IUPAC Name 4-chloro-1H-pyrazole
SMILES C1=C(C=NN1)Cl

Specifications

Melting Point 71.0°C to 73.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 102.52
Percent Purity 98%
Chemical Name or Material 4-Chloropyrazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only