missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloropropiophenone, 98%

Catalog No. AAA1163314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1163314 25 g
AAA1163322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1163314 Supplier Thermo Scientific Chemicals Supplier No. A1163314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6285-05-8 | C9H9ClO | 168.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6285-05-8
MDL Number MFCD00000626

Specifications

Melting Point 33°C to 36°C
Boiling Point 125°C to 126°C (13 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C7H12O
Quantity 25 g
Beilstein 1100638
Synonym 4'-chloropropiophenone, 1-4-chlorophenyl propan-1-one, p-chloropropiophenone, 4-chloropropiophenone, 1-propanone, 1-4-chlorophenyl, propiophenone, 4'-chloro, propiophenone, p-chloro, ethyl p-chlorophenyl ketone, usaf ek-5296, unii-26ay8wja1eAfficher moins
Solubility Information Insoluble in water
InChI Key PUNRPAWKFTXZIW-UHFFFAOYSA-N
SMILES CCC(CC)(C#C)O
IUPAC Name 3-ethylpent-1-yn-3-ol
Molecular Weight (g/mol) 112.17
PubChem CID 22698
Formula Weight 168.62
Percent Purity 98%
Chemical Name or Material 4'-Chloropropiophenone
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-511-6
RTECSNumber UG9275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only