missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloropropiophenone 98.0+%, TCI America™

Catalog No. p-7125090
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C161625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C161625G Supplier TCI America Supplier No. C161625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6285-05-8
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00000626
InChI Key ADCYRBXQAJXJTD-UHFFFAOYSA-N
Synonym 4'-chloropropiophenone, 1-4-chlorophenyl propan-1-one, p-chloropropiophenone, 4-chloropropiophenone, 1-propanone, 1-4-chlorophenyl, propiophenone, 4'-chloro, propiophenone, p-chloro, ethyl p-chlorophenyl ketone, usaf ek-5296, unii-26ay8wja1e
PubChem CID 22697
IUPAC Name 1-(4-chlorophenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(C=C1)Cl

Specifications

Melting Point 35°C
Color White-Yellow
Boiling Point 152°C
Quantity 25 g
Formula Weight 168.62
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Chloropropiophenone
EINECSNumber (3)-3817
RTECSNumber UG9275000
TSCA Yes
Recommended Storage Refrigerator