missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™

Catalog No. p-2799141
Change view
Click to view available options
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC377408000 800 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC377408000 Supplier Thermo Scientific Chemicals Supplier No. 377408000
Only null left

CAS: 873-77-8 | C6H4BrClMg | 215.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 873-77-8 , 60-29-7
Molecular Formula C6H4BrClMg
Molecular Weight (g/mol) 215.76
MDL Number MFCD00009926
InChI Key CDEMHJCJMMOFMB-UHFFFAOYSA-M
Synonym 4-chlorophenylmagnesium bromide, bromo p-chlorophenyl magnesium, bromo 4-chlorophenyl magnesium, p-chlorophenylmagnesium bromide, 4-chlorophenyl magnesium bromide, pubchem24069, 4-chlorophenylmagnesiumbromide, 4-chlorphenylmagnesium bromide, 4-chloro phenylmagnesiumbromide, 4-chlorophenyhnagnesium bromide, grignard reagent
PubChem CID 101888
IUPAC Name magnesium;chlorobenzene;bromide
SMILES ClC1=CC=C([Mg]Br)C=C1

Specifications

Color Black-Brown to Gray
CAS Min % 22.85, 69.54
CAS Max % 30.46, 77.15
Linear Formula Cl(C6H4)MgBr
Concentration 0.9 to 1.2M
Density 0.8500g/mL
Formula Weight 215.76
Packaging AcroSeal™ Glass Bottle
Quantity 800 mL
Physical Form Solution
Chemical Name or Material 4-Chlorophenylmagnesium bromide, 1M Solution in Diethyl Ether
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Harmful if swallowed. May cause drowsiness or dizziness. Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Extremely flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. Store in a dry place. Store in a closed container. Keep container tightly closed. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Recommended Storage Magnesium salts may crystallize below 20°C

RUO – Research Use Only