missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylacetone 97.0+%, TCI America™

Catalog No. C25265G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25265G 5 g
C252625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25265G Supplier TCI America Supplier No. C25265G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5586-88-9
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00045214
InChI Key WEJRYKSUUFKMBC-UHFFFAOYSA-N
Synonym 4-chlorophenylacetone, 1-4-chlorophenyl acetone, 1-4-chlorophenyl propan-2-one, 1-p-chlorophenyl acetone, 1-4-chloro-phenyl-propan-2-one, p-chlorophenyl acetone, 2-propanone, 1-4-chlorophenyl, p-chlorophenylacetone, 4-chlorophenyl acetone
PubChem CID 79699
IUPAC Name 1-(4-chlorophenyl)propan-2-one
SMILES CC(=O)CC1=CC=C(Cl)C=C1

Specifications

Color Yellow
Boiling Point 216°C
Quantity 5 g
Formula Weight 168.62
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chlorophenylacetone