missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenyl Cyclopropyl Ketone 96.0+%, TCI America™

Catalog No. C168125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C168125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C168125G Supplier TCI America Supplier No. C168125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6640-25-1
Molecular Formula C10H9ClO
Molecular Weight (g/mol) 180.631
MDL Number MFCD00001295
InChI Key OPSFCTBBDIDFJM-UHFFFAOYSA-N
Synonym 4-chlorophenyl cyclopropyl ketone, 4-chlorophenyl cyclopropyl methanone, 4-chlorobenzoyl cyclopropane, 4-chlorobenzoylcyclopropane, methanone, 4-chlorophenyl cyclopropyl, 4-chlorophenylcyclopropylmethanone, pubchem16774, acmc-209nvc, cyclopropyl 4-chlorophenyl ketone
PubChem CID 81148
IUPAC Name (4-chlorophenyl)-cyclopropylmethanone
SMILES C1CC1C(=O)C2=CC=C(C=C2)Cl

Specifications

Quantity 25 g
Formula Weight 180.63
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Chlorophenyl Cyclopropyl Ketone