missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenoxyacetic acid, 98+%

CAS: 122-88-3 | C8H7ClO3 | 186.591 g/mol

Supplier:  Thermo Scientific Chemicals A1306536

 View more versions of this product

Catalog No. AAA1306536


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Chlorophenoxyacetic acid
122-88-3
C8H7ClO3
1211804
SODPIMGUZLOIPE-UHFFFAOYSA-N
2-(4-chlorophenoxy)acetic acid
26229
186.59
156°C to 161°C
500 g
MFCD00004305
4-chlorophenoxyacetic acid, 2-4-chlorophenoxy acetic acid, 4-chlorophenoxy acetic acid, 4-cpa, sure-set, tomatotone, tomato fix, acetic acid, 4-chlorophenoxy, p-chlorophenoxyacetic acid, tomato hold
C1=CC(=CC=C1OCC(=O)O)Cl
186.591
CHEBI:1808
≥98%

Chemical Identifiers

122-88-3
186.591
SODPIMGUZLOIPE-UHFFFAOYSA-N
26229
2-(4-chlorophenoxy)acetic acid
C8H7ClO3
MFCD00004305
4-chlorophenoxyacetic acid, 2-4-chlorophenoxy acetic acid, 4-chlorophenoxy acetic acid, 4-cpa, sure-set, tomatotone, tomato fix, acetic acid, 4-chlorophenoxy, p-chlorophenoxyacetic acid, tomato hold
CHEBI:1808
C1=CC(=CC=C1OCC(=O)O)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 204-581-3

RTECSNumber : AG0175000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only