missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenoxyacetic acid, 98+%

Catalog No. p-7020939
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1306522 100 g
AAA1306536 500 g
AAA130650E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1306522 Supplier Thermo Scientific Chemicals Supplier No. A1306522
Only null left

CAS: 122-88-3 | C8H7ClO3 | 186.591 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122-88-3
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00004305
InChI Key SODPIMGUZLOIPE-UHFFFAOYSA-N
Synonym 4-chlorophenoxyacetic acid, 2-4-chlorophenoxy acetic acid, 4-chlorophenoxy acetic acid, 4-cpa, sure-set, tomatotone, tomato fix, acetic acid, 4-chlorophenoxy, p-chlorophenoxyacetic acid, tomato hold
PubChem CID 26229
ChEBI CHEBI:1808
IUPAC Name 2-(4-chlorophenoxy)acetic acid
SMILES C1=CC(=CC=C1OCC(=O)O)Cl

Specifications

Melting Point 156°C to 161°C
Quantity 100 g
Beilstein 1211804
Formula Weight 186.59
Percent Purity ≥98%
Chemical Name or Material 4-Chlorophenoxyacetic acid
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 204-581-3
RTECSNumber AG0175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only