Learn More
4-Chlorophenoxyacetic acid, 98+%
CAS: 122-88-3 | C8H7ClO3 | 186.591 g/mol
Supplier: Thermo Scientific Chemicals A1306522
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Chlorophenoxyacetic acid | |
| 122-88-3 | |
| C8H7ClO3 | |
| 1211804 | |
| SODPIMGUZLOIPE-UHFFFAOYSA-N | |
| 2-(4-chlorophenoxy)acetic acid | |
| 26229 | |
| 186.59 |
| 156°C to 161°C | |
| 100 g | |
| MFCD00004305 | |
| 4-chlorophenoxyacetic acid, 2-4-chlorophenoxy acetic acid, 4-chlorophenoxy acetic acid, 4-cpa, sure-set, tomatotone, tomato fix, acetic acid, 4-chlorophenoxy, p-chlorophenoxyacetic acid, tomato hold | |
| C1=CC(=CC=C1OCC(=O)O)Cl | |
| 186.591 | |
| CHEBI:1808 | |
| ≥98% |
Chemical Identifiers
| 122-88-3 | |
| 186.591 | |
| SODPIMGUZLOIPE-UHFFFAOYSA-N | |
| 26229 | |
| 2-(4-chlorophenoxy)acetic acid |
| C8H7ClO3 | |
| MFCD00004305 | |
| 4-chlorophenoxyacetic acid, 2-4-chlorophenoxy acetic acid, 4-chlorophenoxy acetic acid, 4-cpa, sure-set, tomatotone, tomato fix, acetic acid, 4-chlorophenoxy, p-chlorophenoxyacetic acid, tomato hold | |
| CHEBI:1808 | |
| C1=CC(=CC=C1OCC(=O)O)Cl |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 204-581-3
RTECSNumber : AG0175000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only