missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenol 98.0+%, TCI America™

Catalog No. p-7124307
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
C024325G 25 g
C0243500G 500 g
2 options

Catalog No. C024325G

Supplier: TCI America C024325G

Only null left

Chemical Identifiers

CAS 106-48-9
Molecular Formula C6H5ClO
Molecular Weight (g/mol) 128.555
MDL Number MFCD00002318
InChI Key WXNZTHHGJRFXKQ-UHFFFAOYSA-N
Synonym p-chlorophenol, parachlorophenol, phenol, 4-chloro, 4-hydroxychlorobenzene, phenol, p-chloro, applied 3-78, 4-monochlorophenol, p-chlorophenic acid, p-chlorfenol, 4-chloro-1-hydroxybenzene
PubChem CID 4684
ChEBI CHEBI:28078
IUPAC Name 4-chlorophenol
SMILES C1=CC(=CC=C1O)Cl

Specifications

Color White
Boiling Point 220°C
Quantity 25 g
UN Number 2020
Formula Weight 128.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Chlorophenol
EINECSNumber (3)-0895
RTECSNumber SK2800000
TSCA Yes